Learn More
Biomatik Corporation Guinea pig Endothelin 1 (EDN1) ELISA Kit
Supplier:  Biomatik Corporation EKU03886
Encompass_Preferred
Guinea pig Endothelin 1 (EDN1) ELISA Kit for detecting EDN1 in serum, plasma, tissue homogenates, cell lysates, cell culture supernates, other biological fluids. Species Reactivity: Guinea pig (Cavia porcellus). Detection Range: 6.17-500pg/ml. Sensitivity: 2.82pg/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Accession Number: P97740. Target Synonyms: ET1; PPET1; Preproendothelin-1; Big endothelin-1. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-241-4901
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.