Learn More
Biomatik Corporation Guinea pig Immunoglobulin G2 (IgG2) ELISA Kit
Supplier:  Biomatik Corporation EKU04903
Encompass_Preferred
Guinea pig Immunoglobulin G2 (IgG2) ELISA Kit for detecting IgG2 in serum, plasma, other biological fluids. Species Reactivity: Guinea pig (Cavia porcellus). Detection Range: 308.64-25000ng/ml. Sensitivity: 124.5ng/ml. Format: 96-Well Plate. Assay Type: Competitive Inhibition. Detection Method: Colormetric. Assay Time: 2h. Accession Number: P01862. Target Synonyms: IGHG2; Ig Gamma-2 Chain C Region; Immunoglobulin Heavy Constant Gamma 2; G2m Marker; Immunoglobulin Gm2. Restrictions: For Research Use Only. Not for use in diagnostic procedures.
Catalog No. 50-252-8928
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.