Learn More
Kingfisher Biotech Inc Guinea Pig M-CSF Recombinant Protein
SDP
Supplier:  Kingfisher Biotech Inc RP1655GP005
Encompass
The Guinea Pig M-CSF yeast-derived recombinant protein is not tagged and is naturally endotoxin-free. The purity is greater than 98% as visualized by SDS-PAGE analysis. Guinea Pig M-CSF applications are for cell culture, ELISA standard, and Western Blot Control. Guinea Pig M-CSF yeast-derived recombinant protein can be purchased in multiple sizes. Guinea Pig M-CSF Specifications: (Molecular Weight: 26.1 kDa) (Amino Acid Sequence: EEISEHCSHM IGNGHLQALQ HLIDSQMETS CHITFEFADE EQLKDPVCYL KKAFLLVDDI MEDTMRFRDE TPNAKAIIQL QTLSVRLKSC FTKDYEKQDK ACVRTFYETP LQLMEKIKNV FNETKNLLEE DWKIFTKNCS DSFAKCSSPD VVTKPDCNCL YPKATPSSDL ASVSPHQPFI PSKVPKTGLT WADSEGTEGR SLLPNKQLLH TVDPGTAKHR PPRSTCQSLE (230)) (Gene ID: 100732898). For research use only. Made in the USA
Catalog No. 50-253-2981
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.