Learn More
American Research Products Inc Guinea pig testosterone T EL
Supplier:  American Research Products Inc CSBEQ028156GU
Encompass
Guinea pig testosterone (T) ELISA kit from CUSABIO Detection Range 0 1 ng/mL-20 ng/mL Sensitivity 0 05 ng/mL Sample serum plasma Method Competitive
Catalog No. 50-003-26413