Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ GULP1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5109801
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5109801 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5109801 Supplier Invitrogen™ Supplier No. PA5109801
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MNRAFSRKKD KTWMHTPEAL SKHFIPYNAK FLGSTEVEQP KGTEVVRDAV RKLKFARHIK KSEGQKIPKV ELQISIYGVK ILEPKTKEVQ HNCQLHRISF CADDKTDKRI FTFICKDSES NKHLCYVFDS.

The prompt clearance of cells undergoing apoptosis is critical during embryonic development, normal tissue turnover, inflammation, and autoimmunity. CED6 is an evolutionarily conserved adaptor protein required for efficient engulfment of apoptotic cells by phagocytes.
TRUSTED_SUSTAINABILITY

Specifications

Antigen GULP1
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1.67 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.01% thimerosal; pH 7.3
Gene GULP1
Gene Accession No. Q5PQS4, Q8K2A1, Q9UBP9
Gene Alias 3110030A04Rik; 5730529O06Rik; CED 6; Ced6; CED-6; CED-6 protein; Cell death protein 6 homolog; engulfment adapter protein; FLJ31156; GULP; GULP, engulfment adaptor PTB domain containing 1; GULP1; Gulp-2; OTTHUMP00000205908; OTTHUMP00000205909; OTTHUMP00000205911; Protein GULP; PTB domain adapter protein CED-6; PTB domain adaptor protein CED-6; PTB domain-containing engulfment adapter protein 1
Gene Symbols GULP1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 1-130 of human GULP1 (NP_057399.1).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 314543, 51454, 70676
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.