Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gum arabic, MP Biomedicals™

Catalog No. MP215121890 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 g
500 g
1 kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MP215121890 500 g
ICN15121880 100 g
ICN15121891 1 kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. MP215121890 Supplier MP Biomedicals, Inc Supplier No. 0215121890
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Gum Arabic can be branched polymer of galactose, rhamnose, arabinose, and glucuronic acid as the calcium, magnesium, and potassium salts.

  • It is used in textiles, ceramics, lithography, cosmetics and pharmaceuticals.
  • In the food industry, acacia is mainly used as confectionery, bakery, dairy, drink, and as a microencapsulating agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9000-01-5
MDL Number MFCD00081264

Specifications

Color Off-white
pH 4.50
Physical Form Powder
Quantity 500 g
Moisture ≤15%
Synonym Gum Arabic, Arabicgum, Australiangum, Acacia Syrup
RTECSNumber CE5945000
storageNote1 Store at room temperature (15-30°C).
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.