Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Gum arabic, powder

Catalog No. p-139739
Change view
Click to view available options
Quantity:
250 g
1 kg
2.5 kg
Packaging:
Plastic bottle
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity Packaging
AC258852500 250 g Plastic bottle
AC258850010 1 kg Plastic bottle
AC258850025 2.5 kg Plastic bottle
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. AC258852500 Supplier Thermo Scientific Chemicals Supplier No. 258852500
Only null left
Add to Cart

CAS: 9000-01-5

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 9000-01-5
MDL Number MFCD00081264
Synonym Acacia

Specifications

Color Beige-Yellow to White
Physical Form Powder
Ash Total ash: < 4.0%
Density 1.3500g/mL
Solubility Information Solubility in water: soluble. Other solubilities: insoluble in alcohol
Quantity 250 g
Identification Passes Test (per Eur. Pharm.)
Infrared Spectrum Authentic
Insoluble Matter 0.1% max.
Loss on Drying 15.0% max. (1 g, 105°C)
Packaging Plastic bottle
Specific Gravity 1.35
Chemical Name or Material Gum arabic
Show More Show Less
Hazard Statement GHS H Statement:
May cause respiratory irritation.
Causes skin irritation.
May cause an allergic skin reaction.
Causes serious eye irritation.
May cause allergy or asthma symptoms or breathing difficulties if inhaled.
Precautionary Statement GHS P Statement:
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF INHALED: If breathing is difficult, remove to fresh air and keep at rest in a position comfortable for breathing.
If experiencing respiratory symptoms: Call a POISON CENTER or doctor/physician.
Wear eye protection/face protection.
Avoid breathing dust/fume/gas/mist/vapors/spray.
IF ON SKIN: Wash with plenty of soap and water.
Wear protective gloves/protective clothing/eye protection/face protection.
IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.

WARNING: The information provided on this web site was developed in compliance with European Union (EU) regulations and is correct to the best of our knowledge, information and belief at the date of its publication. The information given is designed only as a guide for safe handling and use. It is not to be considered as either a warranty or quality specification.
EINECSNumber 232-519-5
RTECSNumber CE5945000
TSCA TSCA

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.