Learn More
Cayman Chemical GunosIn 5-dIphosphatesod 5mg
SDP
Supplier:  Cayman Chemical 317045 MG
Encompass_Preferred
A T-type calcium channel blocker inhibits inactivated-state Cav3 2 channels (IC50 2 3 M) and inactivated-state Cav3 1 and Cav3 3 channels by 50 and 39 respectively at 10 M selectively inhibits inactivated-state T-type calcium currents over resting-state currents in rat DRG neurons (IC50s 7 6 and 30 M respectively) reverses the hind limb grip force deficit in a rat model of MIA-induced osteoarthritic pain (ED50 2 mg/kg) dose-dependently increases the paw withdrawal threshold in rat SNL and CCI models of neuropathic pain attenuates cold allodynia in the CCI rat model at 3 mg/kg
Catalog No. 50-299-0701
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.