Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Guvacine hydrochloride, R&D Systems™
SDP

Catalog No. 0234100 Shop All Novus Products
Change view
Click to view available options
Quantity:
100 mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
02-341-00 100 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 02-341-00 Supplier R&D Systems Supplier No. 0234/100

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

A Tocris product, part of R&D Systems. Specific GABA uptake inhibitor

Specific GABA uptake inhibitor. IC50 values are 14, 58, 119 and 1870 μM for hGAT-1, rGAT-2, hGAT-3 and hBGT-1 respectively.

Chemical Identifiers

CAS 6027-91-4
Molecular Formula C6H10ClNO2
Molecular Weight (g/mol) 163.601
InChI Key FGNUNVVTHHKDAM-UHFFFAOYSA-N
Synonym guvacine hydrochloride, 1,2,5,6-tetrahydropyridine-3-carboxylic acid hydrochloride, 1,2,5,6-tetrahydro-pyridine-3-carboxylic acid hydrochloride, guvacine hcl, spectrum1502126, guvacine hydrochloride nmr , solid, 1,2,5,6-tetrahydronicotinic acid hydrochloride, guvacine hydrochloride \ gaba uptak, 1,2,5,6-terahydropyridine-3-carboxylic acid hydrochloride, 1,2,5,6-tetrahydro-3-pyridinecarboxylic acid, hydrochloride salt
PubChem CID 11957555
IUPAC Name 1,2,3,6-tetrahydropyridine-5-carboxylic acid;hydrochloride
SMILES C1CNCC(=C1)C(=O)O.Cl

Specifications

Quantity 100 mg
Formula Weight 163.6
Percent Purity >99%
Product Line Tocris
Chemical Name or Material Guvacine hydrochloride
Recommended Storage Store at Room Temperature
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.