Learn More
Gold Biotechnology Inc GV3101 Agrobacterium Electrocompetent Cells 5x50 µL
Supplier: Gold Biotechnology Inc CC2075X50C
GoldBio’s GV3101 Agrobacterium Electrocompetent Cells allow you to obtain high transformation efficiency in applications such as gDNA or cDNA library construction. Our GV3101 strain harbors the C58 chromosomal backbone containing rifampicin resistance and the Ti plasmid pMP90 (pTiC58DT-DNA) harboring the gentamicin resistance. A functional T-DNA binary system can be built using our GV3101 strains as the T-DNA region has been deleted from the Ti plasmid and instead has a binary vector containing the missing T-region. The binary system makes possible to transfer genetic material into a host plant’s genome. Our system is often used for Agrobacterium-mediated transformation in mono and dicotyledonous species such as Arabidopsis thaliana, tobacco, potato, soybeans and corn.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.