Learn More
TARGETMOL CHEMICALS INC GW-6604 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T99475MG
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. GW-6604 is an ALK5 inhibitor and shows clear antifibrotic effects resulting in liver function improvement. purity: 99%
Catalog No. 50-257-6185
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.