Learn More
Selleck Chemical LLC GW2580 25mg 870483-87-7 SC-203877
SDP
Supplier:  Selleck Chemical LLC S804225MG
Encompass_Preferred
GW2580 (SC-203877) is a selective CSF-1R inhibitor for c-FMS with IC50 of 30 nM, 150- to 500-fold selective compared to b-Raf, CDK4, c-KIT, c-SRC, EGFR, ERBB2/4, ERK2, FLT-3, GSK3, ITK, JAK2 etc. *For Research & Development use only.  Product is not intended for drug, household, or human consumption.
Catalog No. 50-136-5601