Learn More
Selleck Chemical LLC GW441756 S2891-1g
SDP
Supplier:  Selleck Chemical LLC S28911G
Encompass_Preferred
GW441756 is a potent selective inhibitor of TrkA with IC50 of 2 nM with very little activity to c-Raf1 and CDK2 GW441756 produces a relevant increase of caspase-3 that leads to apoptosis
Catalog No. 50-311-1661
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.