Learn More
AdipoGen GW501516 (1 mg)
Supplier: AdipoGen AGCR13641M001
Chemical. CAS: 317318-70-0. Formula: C21H18F3NO3S2. MW: 453.5. Potent and selective peroxisome proliferator-activated receptor (PPAR) delta agonist/activator with high affinity (Ki=1nM) and potency (EC50=1nM) for PPARdelta and >1000 fold selectivity over PPARalpha and PPARgamma. Increases ABC A1 transporter expression and induces apolipoprotein A1-mediated cholesterol efflux in vitro. Increase serum HDL cholesterol and lowers small-dense LDL levels in obesity in vivo models. Reduces plasma triglyceride concentration. Increases expression of genes involved in lipid utilization, beta-oxidation, cholesterol efflux and energy uncoupling, as well as apolipoprotein-A1 specific efflux of intracellular cholesterol, indicating the muscle tissue is an important target. Model compound for a new type of obesity therapeutic, as well as a selective pharmacological tool for understanding lipid metabolism.
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.