Learn More
Abcam GW7647, PPARalpha agonist, 10MG
Supplier: Abcam AB14112410MG
MW 502.8 Da, Purity >99%. Highly potent, selective PPARα agonist (EC₅₀ values are 6, 1100 and 6200 nM for PPARα, PPARγ and PPARδ respectively). Pharmacologically active in vivo.
The product is subject to the following: Abcam Restricted Use Statement