Learn More
Cayman Chemical GW 9662, 22978-25-2, 5 mg
SDP
Supplier:  Cayman Chemical 707855
Encompass_Preferred
Molecular Formula C13H9ClN2O3, Purity ≥98%, Formula Weight 276.7, Synonyms: 2-chloro-5-nitrobenzanilide; The peroxisome proliferator-activated receptor (PPAR) is the nuclear receptor responsible for transducing the therapeutic activity of the thiazolidinediones. Thiazolidinediones are a group of structurally related synthetic PPAR agonists with antidiabetic actions in vivo. Rosiglitazone (BRL 49653) is a prototypical thiazolidinedione and has served as a reference compound for this class. There are many PPAR agonists, including 15-deoxy-.DELTA.12,14-prostaglandin J2 and azelaoyl PAF, which are naturally derived. However, only a few antagonists have been reported. GW 9662 blocks the PPAR-induced differentiation of monocytes to osteoclasts by >90% at a dose of 0.1 M. It is therefore a much more potent antagonist than BADGE, which is another reported PPAR antagonist.
Catalog No. NC9988826
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.