Learn More
TARGETMOL CHEMICALS INC GYPENOSIDE XVII 10MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T3385
Encompass_Preferred
Also available in 1 mg 5 mg 25 mg 50 mg 100 mg and bulk. Please contact Fisher for quotes. Gypenoside XVII (Gynosaponin S) confers protection against A(beta)25-35-induced neurotoxicity through estrogen receptor-dependent activation of PI3K/Akt pathways activation of Nrf2/ARE/HO-1 and inactivation of GSK-3(beta) pathways. purity: 99%
Catalog No. 50-258-0289
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.