Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
MP Biomedicals Gypsy Moth Diet Without Agar
Previously been known as the high wheat germ diet.
$63.50 - $85.00
Specifications
Product Type | Animal Diet |
---|---|
Autoclavable | Not Autoclavable |
Disposable | Single-use |
Sterility | Non-sterile |
Storage Requirements | Room Temperature |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
MP296029401
|
MP Biomedicals
0296029401 |
1 L |
Each for $63.50
|
|
|||||
MP296029404
|
MP Biomedicals
0296029404 |
4 L |
Each for $85.00
|
|
|||||
Description
- Available with agar included, packaged separately or omitted completely
- Methyl paraben and sorbic acid added as preservatives
Specifications
Animal Diet | |
Single-use | |
Room Temperature | |
0.2286 m |
Not Autoclavable | |
Non-sterile | |
9 in. |
Unless specified otherwise, MP Biomedical's products are for research or further manufacturing use only, not for direct human use.
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title