Learn More
TARGETMOL CHEMICALS INC H-1152 DIHYDROCHLORIDE 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T35328
Encompass_Preferred
Also available in 1 mg 2 mg 10 mg 25 mg 50 mg 100 mg 500 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. H-1152 dihydrochloride (H-1152 2HCl) is a specific inhibitor of Rho-associated protein kinase (ROCK) with an IC50 of 12 nM and a Ki of 1.6 nM. H-1152 dihydrochloride inhibits PKA PKC PKG Aurora A and CaMKII with IC50 values of 3.03 uM 5.68 uM 0.360 uM 0.745 uM and 0.180 uM respectively. purity: 98%
Catalog No. 50-258-0226
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.