Learn More
IKA™ H 135.10 Square Carrier without Handle
SDP
Catalog No. 01183253 Shop All IKA Works Products
Change view
Click to view available options
For Use With (Equipment):
Variety of stirrers
Catalog No. For Use With (Equipment)
01-183-253 Variety of stirrers
1 options
This item is not returnable. View return policy
Catalog No. 01-183-253 Supplier IKA™ Supplier No. 25000832
Add to Cart
Add to Cart
This item is not returnable. View return policy
Square, hollow, anodized aluminum carrier suitable for a variety of stir plates.
  • Hollow, anodized aluminum construction
  • Square shape
  • Temperature resistant
  • Suitable for use with a variety of stirrers:
    • IKA Plate (RCT Digital)
    • C-MAG HS 7 Control
    • RCT 5 Digital
    • RET Control-visc
    • RET Control-visc, White
    • RH Basic
    • RH Basic, White
    • RH Digital
    • RH Digital, White
    • RCT Basic
    • RET Basic
Specifications
For Use With (Equipment) Variety of stirrers
Shape Square
Temperature (Metric) 300°C