Learn More
Invitrogen™ H-2Db Monoclonal Antibody (27-11-13S), FITC

Description
H-2Db Monoclonal Antibody for Flow
Specifications
Specifications
| Antigen | H-2Db |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Clone | 27-11-13S |
| Concentration | 0.5 mg/mL |
| Conjugate | FITC |
| Formulation | PBS with <0.1% sodium azide; pH 7.4 |
| Gene | H2-D1 |
| Gene Accession No. | P01895, P01896, P01897, P01899, P01900, P14426, P14427 |
| Gene Alias | H-2 class I histocompatibility antigen, D-B alpha chain; H-2D; H2-D; H-2D cell surface glycoprotein; H-2D(B); H2D1; H2-D1; H2DB; H2-DC1-beta transplantation antigen protein; H2L; H2LD; histocompatibility 2, D region locus 1; MHC class I antigen; MHC class I H2 antigen; MHC class I H-2Dp; MHC H2-D-q alpha-chain; Q5k protein |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.