Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ H-2Db Monoclonal Antibody (28-14-8), FITC
GREENER_CHOICE

Catalog No. 501137715
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-113-7715 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-113-7715 Supplier Invitrogen™ Supplier No. A15442
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

H-2Db Monoclonal Antibody for Flow

This protein is an MHC class I alloantigen, which is involved in antigen presentation to T cells expressing CD3/TCR and CD8. HLA and MHC antibodies play a significant role in Immunopeptidomics, facilitating the identification and characterization of neoantigens through high-performance liquid chromatography coupled to tandem Mass Spectrometry.
TRUSTED_SUSTAINABILITY

Specifications

Antigen H-2Db
Applications Flow Cytometry
Classification Monoclonal
Clone 28-14-8
Concentration 0.5 mg/mL
Conjugate FITC
Formulation PBS with <0.1% sodium azide; pH 7.4
Gene H2-D1
Gene Accession No. P01895, P01896, P01897, P01899, P01900, P14426, P14427
Gene Alias H-2 class I histocompatibility antigen, D-B alpha chain; H-2D; H2-D; H-2D cell surface glycoprotein; H-2D(B); H2D1; H2-D1; H2DB; H2-DC1-beta transplantation antigen protein; H2L; H2LD; histocompatibility 2, D region locus 1; MHC class I antigen; MHC class I H2 antigen; MHC class I H-2Dp; MHC H2-D-q alpha-chain; Q5k protein
Gene Symbols H2-D1
Host Species Mouse
Immunogen C3H.SW splenocytes (Mouse).
Purification Method Purified
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 14964
Target Species Mouse
Content And Storage 4°C, store in dark
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.