Learn More
AdipoGen H-89 . dihydrochloride,Chemical. CAS: 130964-39-5
SDP
Supplier:  AdipoGen AGCR10002M005
Encompass_Preferred
H-89 . dihydrochloride, AdipoGen Life Sciences. Chemical. CAS: 130964-39-5 | 127243-85-0 (free base). Formula: C20H20BrN3O2S . 2HCl. MW: 446.4 . 72.9. Cell permeable potent and selective cAMP- and cGMP-dependent protein kinase (PKA and PKG) inhibitor. Protein kinase C? (PKC?) inhibitor. Ca2+/calmodulin-dependent protein kinase II inhibitor. Casein kinase I inhibitor. Myosin light chain kinase (MLCK) inhibitor. Apoptosis enhancer. Tool to study protein crystal structure-inhibitor interactions. Rho kinase inhibitor. Cell proliferation inhibitor. Review.
Catalog No. 50-464-530
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.