Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

H-B Instrument™ Durac™ Salt Brine Plain Form Hydrometers

Choose from two plain form models with ranges from 0 to 100% or 0 to 26.5% to measure sodium chloride saturation of salt brines

$117.80 - $130.00

Specifications

Certifications/Compliance Traceable to NIST
Content And Storage Salt Brine hydrometers should be stored in a clean, dry, safe place, and should not be subjected to extreme temperatures.
For Use With (Application) Used in various industries including food, fishing, and transportation.
View More Specs

Products 2
Catalog Number Mfr. No. Type Specific Gravity Range Accuracy Increments Price Quantity & Availability  
Catalog Number Mfr. No. Type Specific Gravity Range Accuracy Increments Price Quantity & Availability  
13-202-433
View Documents
H-B Instrument™
B618090100
Hydrometer (% by Weight) 0 to 26.5% ±0.5% 0.5%
Each for $117.80
Only null left
Add to Cart
 
13-202-655
View Documents
H-B Instrument™
B618090000RMD
Hydrometer (% by Saturation) 0 to 100% ±1% 1%
Each for $130.00
Only null left
Add to Cart
 
Description

Description

  • Lead-free ballast and glass
  • Individually serialized
  • Temperature standardization: 60°F (15°C)

Order Info

Should you require an individual Hydrometer Calibration Report, factory calibrated hydrometers or custom calibration services are available.

Certifications

Packaged with a multi-language (English, French, German, Italian, Portuguese, and Spanish) Statement of Accuracy indicating accuracy traceable to NIST. Hand inspected by U.S. quality team.

Specifications

Specifications

Traceable to NIST
Salt Brine hydrometers should be stored in a clean, dry, safe place, and should not be subjected to extreme temperatures.
Used in various industries including food, fishing, and transportation.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.