Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

H-B Instrument™ Easy-Read™ Individually Calibrated Liquid-In-Glass Thermometers: Partial Immersion
GREENER_CHOICE

Catalog No. 13202872 Shop All H-B Instrument Company Products
Change view
Click to view available options
Temperature Range (Metric):
-10°C to +260°C
-10°C to +70°C
-20°C to +110°C
-20°C to +150°C
Calibration Points:
0, 100, 200°C
0, 37, 56°C
0, 37, 56, 100, 230, 250°C
0, 4, 20, 37, 44.5, 103°C
0, 50, 100°C
0, 60, 120°C
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Temperature Range (Metric) Calibration Points
13-202-872 -10°C to +260°C 0, 100, 200°C
13-201-476 -20°C to +110°C 0, 50, 100°C
13-201-477 -20°C to +110°C 0, 4, 20, 37, 44.5, 103°C
13-201-563 -20°C to +150°C 0, 60, 120°C
13-201-561 -10°C to +70°C 0, 37, 56°C
13-202-528 -20°C to +110°C 0, 37, 56°C
13-202-873 -10°C to +260°C 0, 37, 56, 100, 230, 250°C
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 13-202-872 Supplier H-B Instrument™ Supplier No. B602021300
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Individual Calibration Report as needed for many standard operating procedures

  • Fill: Black spirit
  • Each thermometer is individually calibrated
  • Celsius-scale
  • Designed to be very easy to read
  • Replace mercury thermometers with the same accuracy without any hazards
  • Nontoxic, nonhazardous, biodegradable
  • NIST-Certified detailed, laminated report of calibration
  • Each thermometer features a black liquid and yellow back glass which creates a distinct contrast making this thermometer very easy to read
  • Individually calibrated to NIST standards at specific temperature and supplied with NIST report of calibration stating corrections to one-tenth the smallest division
  • Contain an environmentally friendly non-mercury liquid that is non-toxic, non-hazardous, and fully biodegradable
  • In case of breakage the liquid can be cleaned up easily with soap and water

  • “Lead-free”, yellow back glass and black liquid provides easy readability and fewer reading errors
  • Contains non-toxic, biodegradable, Enviro-Safe™ liquid
  • EnviroKleen™ certified

Applications:

Ideal for use in chemical/petrochemical laboratories, universities, scientific institutions, food and beverage processing, pharmaceutical and medical industries, dairies, breweries and distilleries.

Certifications

Each thermometer includes an individualized Thermometer Calibration Report stating corrections to one-tenth of the smallest division, unique report number and NIST™ standards used for the calibration. Individually serialized and made in the USA.

Specifications

Calibration Points 0, 100, 200°C
Certifications/Compliance Each thermometer includes an individualized Thermometer Calibration Report stating corrections to one-tenth of the smallest division, unique report number and NIST standards used for the calibration. Individually serialized and made in the USA
For Use With (Application) Ideal for use in chemical/petrochemical laboratories, universities, scientific institutions, food and beverage processing, pharmaceutical and medical industries, dairies, breweries and distilleries.
Grade Precision
Immersion Type Partial Immersion
Length (English) Overall 14 in.
Length (Metric) 350 mm
Length (Metric) Overall 350 mm
Material Glass
Thermometer Fill Enviro-Safe™ Liquid
Product Type Individually Calibrated Partial Immersion Liquid-In-Glass Thermometers
Max. Temperature (English) 500°F
Max. Temperature (Metric) 260°C
Min. Temperature (English) 14°F
Min. Temperature (Metric) -10°C
Temperature Range (Metric) -10°C to +260°C
Calibrated Yes
Accuracy ±1°C; ±1.5°C above 110°C, ±2°C above 200°C
Subdivision 1°C
Immersion Length 76 mm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.