Learn More
Gentarget H CD8A EXPRESSION LENTIVIRUS
Supplier:  Gentarget LVP1151
Encompass
This Pre-made lentiviral particles expresses human target, CD8a (CD8a molecule), Alternative Names: CD8; Leu2; MAL; p32. The sub-cloned codon sequence is identical (100% match) to CDS region in NCBI ID: NM_001768.6
Catalog No. NC1955123
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.