Learn More
Thermo Scientific™ H-Flex 1 Swinging Bucket Rotor and Accessories
Description
Spin more with less! Whether clinical and diagnostic applications or cell culture and other research, now spin conical tubes, blood tubes and microplates all in one rotor for a small-footprint General Purpose centrifuge. High capacity and flexibility—all in a small footprint.
- Auto-Lock and Click Seal biocontainment lids enable fast, one-handed rotor exchanges and lid locking
- Spins blood tubes, conical tubes and microplates without changing rotor or buckets
- Get results faster without the need to change hardware
- Limit rotor maintenance and contamination potential with slide-coat technology eliminating the need for grease and lubricants
Specifications
Specifications
| Description | 400 mL Bottle/Centrilab Adapter |
| Material | Molded Polyamide Nylone |
| Type | Swinging Bucket Rotor |
| Capacity | 1 x 400 mL Bottle or Centrilab Each Adapter |
| Max. Speed | 4297 |
| For Use With (Equipment) | Thermo Scientific Sorvall Legend X1 and ST16 Series, Thermo Scientific Multifuge X1 and Megafuge 16 Series, Thermo Scientific SL 16 Series |
| Biocontainment | Yes |
| Autoclavable | Autoclavable |
| Quantity | Set of 2 |