Learn More
Description
- Human on Human Immunodetection Kit is designed for the detection of human primary antibodies on human tissue samples.
- Tested Applications: IHC, WB, ICC/IF
- Species Reactivity: Human
Specifications
Specifications
| Product Type | Immunodetection Kit |
| Target Species | Human |
| Quantity | 1 Kit |
| For Use With (Application) | Immunohistochemistry (IHC), Immunocytochemistry/Immunofluorescence, Immunohistochemistry (Paraffin) |
| Product Line | Novus |
| Species | Human |
| Target Specificity | H.O.H.(TM) (Human on Human) |
| Content And Storage | Store at 4°C. |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
