Learn More
Bachem H-Phe-Arg-bNA · 2 HCl
SDP
Supplier:  Bachem 4000885.0250
Encompass_Preferred
H-Phe-Arg-bNA · 2 HCl, 100929-99-5, C25H30N6O2 · 2 HCl, Mr 519.47, 250mg. MC-207110, PAbN. Substrate for dipeptidyl aminopeptidase I (cathepsin C). A broad-spectrum efflux pump inhibitor in gram-negative bacteria. PAbN has been evaluated as an adjunct to antibiotics, as efflux in bacteria mediates antibiotic resistance. Vecchione et al. demonstrated that Phe-Arg-b-naphthylamide could potentiate drug activity also against a gram-positive bacterium. PAbN permeabilizes the outer membrane of gram-negative bacteria (Lamers et al.).
Catalog No. 50-396-868
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.