Learn More
Bachem H-Phe-Met-Arg-Phe-NH2
SDP
Supplier:  Bachem 4001566.0025
Encompass_Preferred
H-Phe-Met-Arg-Phe-NH2 Acetate, 152165-14-5, C29H42N8O4S, Mr 598.77, 25mg. Molluscan Cardioexcitatory Neuropeptide, FMRFamide, FMRF-amide. A molluscan neuropeptide isolated from the ganglia of Macrocallista nimbosa. FMRF-amide seems to be involved in neuronal NO production in gastropoda. CAS Number (net): 64190-70-1.
Catalog No. 50-259-679