Learn More
Bachem H-Pro-Gly-Pro-OH
SDP
Supplier:  Bachem 4035991.0005
Encompass_Preferred
H-Pro-Gly-Pro-OH, 7561-51-5, C12H19N3O4, Mr 269.30, 5mg. H-Pro-Gly-Pro-OH (PGP) is a tripeptide generated from the breakdown of extracellular matrix collagen and is specifically chemotactic for neutrophils in vitro and in vivo. Ac-PGP generated by N-terminal acetylation of PGP can enhance this chemotactic potential. PGP was identified as physiological substrate for the aminopeptidase activity of leukotriene A4 hydrolase. PGP is a biomarker for chronic obstructive pulmonary disease (COPD).
Catalog No. 50-383-355
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.