Learn More
Cliawaived Inc CLIA waived H. pylori Test Kit
SDP
Catalog No. NC1859816
Encompass_Preferred
(25 Tests)A rapid qualitative immunoassay intended to detect the presence of IgG antibodies specific to Helicobacter pylori (H. pylori) in human whole blood. FDA cleared and CLIA waived. (FDA 510K#: K024360)
This item is not returnable. View return policy
Catalog No. NC1859816 Supplier Cliawaived Inc Supplier No. CLIA059476
(25 Tests)A rapid qualitative immunoassay intended to detect the presence of IgG antibodies specific to Helicobacter pylori (H. pylori) in human whole blood. FDA cleared and CLIA waived. (FDA 510K#: K024360)