Learn More
CPC Scientific H-Ser-Gln-Asn-Phe-psi(CH2NH)Pro-Ile-Val-Gln-OH (trifluoroacetate salt) 0.5MG
SDP
Supplier:  CPC Scientific HIV023A
Encompass
SQNF-psi[CH2NH]PIVQ represents a competitive inhibitor of the HIV-1 protease (Ki = 1 ?M). The coupling of this affinity ligand to agarose resin enabled the rapid purification of HIV-1 protease.ONE-LETTER SEQUENCE: SQNF-psi[CH2NH]PIVQMOLECULAR FORMULA: C42H67N11O12MOLECULAR WEIGHT:918.06STORAGE CONDITIONS: -20 5C, CAS REGISTRY NUMBER: [124869-92-7], RESEARCH AREA: AntimicrobialREFERENCES: J.C.Heimbach et al., Biochem. Biophys. Res. Commun., 164, 955 (1989)
Catalog No. 50-210-9573
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.