Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

H-Ser-Gly-OH, MP Biomedicals™

Catalog No. ICN15660880 Shop All MP Biomedicals, Inc Products
Change view
Click to view available options
Quantity:
100 mg
250 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
ICN15660880 100 mg
ICN15660883 250 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. ICN15660880 Supplier MP Biomedicals, Inc Supplier No. 0215660880

Chemical Identifiers

CAS 687-63-8
Molecular Formula C5H10N2O4
Molecular Weight (g/mol) 162.145
MDL Number MFCD00037778
InChI Key WOUIMBGNEUWXQG-VKHMYHEASA-N
Synonym h-ser-gly-oh, l-serylglycine, ser-gly, serylglycine, chembl90610, serinyl-glycine, l-ser-gly, ser-gly-oh, glycine, l-seryl, s-2-2-amino-3-hydroxypropanamido acetic acid
PubChem CID 7009644
ChEBI CHEBI:74814
IUPAC Name 2-[[(2S)-2-amino-3-hydroxypropanoyl]amino]acetic acid
SMILES C(C(C(=O)NCC(=O)O)N)O

Specifications

Color White
Quantity 100 mg
Physical Form Powder
Assay Percent Range >99%
Formula Weight 162.14
Recommended Storage Store at room temperature (15 to 30°C).
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.