Learn More
Bachem (Thr1)-PAR-1 (1-5) amide (human) Trifluoroacetate
SDP
Supplier:  Bachem 4033683.0001
Encompass_Preferred
(Thr1)-PAR-1 (1-5) amide (human) Trifluoroacetate, H-Thr-Phe-Leu-Leu-Arg-NH2, 197794-83-5, C31H53N9O6, Mr 647.82, 1mg. (Thr1)-TRAP-5 amide, TFLLRamide, TFLLR-NH2 Protease-Activated Receptor 1 (PAR1) Agonist. TFLLR-amide, a selective agonist of PAR-1.
Catalog No. 50-383-131