Learn More
Bachem H-Tyr-Tyr-Tyr-OH
SDP
Supplier:  Bachem 4005242.0250
Encompass_Preferred
H-Tyr-Tyr-Tyr-OH, 7390-78-5, C27H29N3O7, Mr 507.54, 250mg. The bitter taste of tri-tyrosine, YYY, is due to its marked hydrophobicity. Narai-Kanayama and Aso showed that Tyr-Tyr (G-3480) and Tyr-Tyr-Tyr efficiently inhibit angiotensin I-converting enzyme (ACE) from rabbit lung, their IC50 values were 34 μM and 51 μM, respectively. These two tyrosine peptides exhibited a mix of competitive and noncompetitive inhibitions.
Catalog No. 50-259-926
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.