Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
CERILLIANT™ Heroin Solution, 1.0 mg/mL in Acetonitrile, Sold by MilliporeSigma™ Supelco™
Certified reference material
Supplier: MilliporeSigma Supelco H0381ML
Description
Heroin, or diacetylmorphine, is an opiate analgesic most commonly used as an illcit recreational drug. Street heroin is considered to be one of the most harmful recreational drugs due to the use of adulterants and the risk of contracting disease from intravenous use. This drug reference standard is suitable for use in GC/MS or LC/MS heroin testing methods for applications in clinical toxicology, pain prescription monitoring, forensic analysis, and urine drug testing.
Specifications
| 75-05-8, 561-27-3 | |
| MFCD00468072 | |
| 1.0 mg/mL in acetonitrile | |
| Ampule | |
| 2°C (35.6°F) - closed cup |
| C21H23NO5 | |
| UN 1648 3 / PGII | |
| 369.41 | |
| 1 mL |
Safety and Handling
H332-H319
H312
Recommended Storage : -20°C
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
CERILLIANT is a registered trademark of Cerilliant Corporation
Spot an opportunity for improvement?Share a Content Correction