Learn More
Bulbman Inc Sylvania 64150 T2.5 Halogen Lamp, 55 Watt, 12 Volt
SDP
Supplier:  Bulbman Inc 101694
Encompass_Preferred
Sylvania 64150 T2.5 halogen lamp operates at 55 watts and 12 volts. It has a P14.5s base, 1621 initial lumens, and an average life of 225 hours.
Catalog No. NC0820808
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.