Learn More
Sino Biological H10N3 HA PAb 100UL 11693-RP02-100
Supplier:  Sino Biological 11693RP02100
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant H10N3 (A/duck/Hong Kong/786/1979) HA / Hemagglutinin (Catalog#11693-V08H; BAF46762.1; Met1-Ile525). H10N3 (A/duck/Hong Kong/786/1979) HA / Hemagglutinin specific IgG was purified by H10N3 (A/duck/Hong Kong/786/1979) HA / Hemagglutinin affinity chromatography.
Catalog No. 50-241-1758
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.