Learn More
Invitrogen™ H1FX Polyclonal Antibody

Description
Gently mix before use. Optimal concentrations and conditions for each application should be determined by the user. Immunogen sequence: AHKAKKAAPG AAGSRRADKK PARGQKPEQR SHKKGAGAKK DKGGKA Highest antigen sequence indentity to the following orthologs: Mouse - 39%, Rat - 39%.
Specifications
Specifications
| Antigen | H1FX |
| Applications | Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Concentration | 0.1 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 |
| Gene | H1-10 |
| Gene Accession No. | Q92522 |
| Gene Alias | H1 histone family member X; H1 histone family, member X; H1.10; H1FX; H1X; Histone H1.X; histone H1x |
| Gene Symbols | H1-10 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.