Learn More
Sino Biological Influenza A H1N1 (A/swine/Guangxi/NS2176/2012) Neuraminidase / NA (His Tag) 100ug
Supplier:  Sino Biological 40735V07H100
Encompass_Preferred
A DNA sequence encoding the influenza A virus (A/swine/Guangxi/NS2176/2012 (H1N1)) neuraminidase (AIE51967) (His36-Lys469), termed as NA, was fused with a N-terminal polyhistidine tag.
Catalog No. 50-255-3613
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.