Learn More
Sino Biological Influenza A H1N1 Neuraminidase / NA (N295S mutation) (Active) 800U
Supplier:  Sino Biological 11058VNAHC2800
Encompass_Preferred
A DNA sequence encoding the Influenza A virus (A/California/04/2009 (H1N1)) neuraminidase (ACP41107.1) (Met 1-Lys 469) was expressed, the cell lysates are collected, and bio-activity was tested. There is an amino acid change from Asparagine to Serine (N295S mutation) in NA / Neuraminidase.
Catalog No. 50-255-4905
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.