Learn More
Creative Biomart Recombinant Dengue-2 Antigen, His-tagged
Supplier:  Creative Biomart H1V1305
Encompass_Preferred
The Recombinant Dengue 2 Envelope Hydrophobic Domain E. coli derived contains 225 amino acids (228-485) from dengue 2 envelope. The protein is fused to a His tag at C-terminal and purified by standard
Catalog No. 50-208-4683
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.