Learn More
Sino Biological H1N2 HA PAb 20UL 11703-RP02-20
Supplier:  Sino Biological 11703RP0220
Encompass_Preferred
Produced in rabbits immunized with purified, recombinant H1N2 (A/swine/Guangxi/13/2006) HA / Hemagglutinin (Catalog#11703-V08H; ABQ42444.1; Met 1-Gln 529). H1N2 (A/swine/Guangxi/13/2006) HA / Hemagglutinin specific IgG was purified by H1N2 (A/swine/Guangxi/13/2006) HA / Hemagglutinin affinity chromatography.
Catalog No. 50-240-7668
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.