Learn More
Molecular Devices CellXpress .ai pipette tips, 1000ul wide bore 1.2mm with filter, conductive tip, case of 3840 tips (40 boxes of 96 tips)- Sterile
Supplier:  Molecular Devices YY000350
Encompass_Preferred
CellXpress .ai pipette tips, 1000ul wide bore 1.2mm with filter, conductive tip, case of 3840 tips (40 boxes of 96 tips)- Sterile
Catalog No. 50-000-14500
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.