Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

H2L 5765834, Tocris Bioscience™

Catalog No. 487050
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
48-705-0 50 mg
48-701-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 48-705-0 Supplier R&D Systems Supplier No. 4870/50
This item is not returnable. View return policy

LPA1,3,5 antagonist

Antagonist of the lysophosphatidic acid receptors LPA1, LPA5 and LPA3 (IC50 values are 94, 463 and 752 nM respectively). Exhibits no effect at LPA2 or LPA4 receptors.

Chemical Identifiers

CAS 420841-84-5
Molecular Formula C21H12N2O7
Molecular Weight (g/mol) 404.334
InChI Key HFYPTENHTPNXGP-UHFFFAOYSA-N
Synonym 2-3-4-nitrophenoxy phenyl-1,3-dioxoisoindole-5-carboxylic acid, 2-3-4-nitrophenoxy phenyl-1,3-dioxoisoindoline-5-carboxylic acid, 2,3-dihydro-2-3-4-nitrophenoxy phenyl-1,3-dioxo-1h-isoindole-5-carboxylic acid, 2-3-4-nitrophenoxy phenyl-1,3-dioxo-2,3-dihydro-1h-isoindole-5-carboxylic acid
PubChem CID 1365686
IUPAC Name 2-[3-(4-nitrophenoxy)phenyl]-1,3-dioxoisoindole-5-carboxylic acid
SMILES C1=CC(=CC(=C1)OC2=CC=C(C=C2)[N+](=O)[O-])N3C(=O)C4=C(C3=O)C=C(C=C4)C(=O)O

Specifications

Quantity 50 mg
Formula Weight 404.33
Percent Purity >99%
Chemical Name or Material H2L 5765834
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.