Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

H2L5186303, Tocris Bioscience™

Catalog No. 487850
Change view
Click to view available options
Quantity:
10 mg
50 mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
48-785-0 50 mg
48-781-0 10 mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Catalog No. 48-785-0 Supplier R&D Systems Supplier No. 4878/50
This item is not returnable. View return policy

Potent and selective LPA2 receptor antagonist

Potent and selective lysophosphatidic acid 2 (LPA2) receptor antagonist (IC50 values are 8.9, 1230 and 27354 nM for LPA2, LPA3 and LPA1 receptors respectively, in a LPA-elicited calcium mobilization assay).

Chemical Identifiers

CAS 139262-76-3
Molecular Formula C26H20N2O8
Molecular Weight (g/mol) 488.452
InChI Key HZFPOTBCYPWQSH-DZDAAMPGSA-N
Synonym z,z-4,4'-1,3-phenylenebis oxy-4,1-phenyleneimino bis 4-oxo-2-butenoic acid, 2z-3-4-3-4-2z-3-carboxyprop-2-enamido phenoxy phenoxy phenyl carbamoyl prop-2-enoic acid
PubChem CID 90488981
IUPAC Name (Z)-4-[4-[3-[4-[[(Z)-3-carboxyprop-2-enoyl]amino]phenoxy]phenoxy]anilino]-4-oxobut-2-enoic acid
SMILES C1=CC(=CC(=C1)OC2=CC=C(C=C2)NC(=O)C=CC(=O)O)OC3=CC=C(C=C3)NC(=O)C=CC(=O)O

Specifications

Quantity 50 mg
Formula Weight 488.45
Percent Purity >98%
Chemical Name or Material H2L5186303
Recommended Storage Store at +4°C
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.