Learn More
TARGETMOL CHEMICALS INC H3B-120 5MG
SDP
Supplier:  TARGETMOL CHEMICALS INC T11529
Encompass_Preferred
Also available in 1 mg 10 mg 25 mg 50 mg 100 mg 1 mL 10 mM (in DMSO) and bulk. Please contact Fisher for quotes. H3B-120 is a highly selective competitive and allosteric carbamoyl phosphate synthetase 1 (CPS1) inhibitor (IC50 1.5 uM; Ki 1.4 uM) with anti-cancer activity. purity: 99%
Catalog No. 50-258-1522
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.