Learn More
Medchemexpress LLC HY-112596 5mg Medchemexpress, H3B-6545 CAS:2052130-80-8 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY1125965MG
Encompass_Preferred
Medchemexpress, HY-112596 5mg H3B-6545 CAS:2052130-80-8 H3B-6545 is an oral, selective estrogen receptor covalent antagonist (SERCA) for the research of metastatic ER-positive, HER2-negative breast cancer. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-0822