Learn More
Medchemexpress LLC HY-112596A 5mg Medchemexpress, H3B-6545 (hydrochloride) CAS:2052132-51-9 Purity:>98%
SDP
Supplier:  Medchemexpress LLC HY112596A5MG
Encompass_Preferred
Medchemexpress, HY-112596A 5mg H3B-6545 (hydrochloride) CAS:2052132-51-9 H3B-6545 hydrochloride is an oral, selective estrogen receptor covalent antagonist (SERCA) for the research of metastatic ER-positive, HER2-negative breast cancer. Purity:>98% Medchemexpress has over 10000 novel life-science reagents, reference compounds, APIs and natural compounds for laboratory and scientific use. Other quantity can also be offered.
Catalog No. 50-208-0823
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.